missing translation for 'onlineSavingsMsg'
Learn More

[(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) Dichloride, TCI America™

Catalog No. B3068250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3068250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3068250MG Supplier TCI America Supplier No. B3068250MG
Only null left

Chemical Identifiers

CAS 134524-84-8
Molecular Formula C44H34Cl2P2Ru
Molecular Weight (g/mol) 796.674
MDL Number MFCD01073794
InChI Key BYFIUAVPOJGEBX-UHFFFAOYSA-N
Synonym Dichloro[(S)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II)
PubChem CID 131664235
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium;dihydrochloride
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.Cl.[Ru]

Specifications

Quantity 250 mg
Formula Weight 794.66
Physical Form Crystalline Powder
Chemical Name or Material [(S)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl]ruthenium(II) Dichloride