missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methoxy-2-propanol 98.0+%, TCI America™

Catalog No. M24081ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24081ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24081ML Supplier TCI America Supplier No. M24081ML

Chemical Identifiers

CAS 26550-55-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD01632588
InChI Key ARXJGSRGQADJSQ-BYPYZUCNSA-N
Synonym s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
PubChem CID 6993459
IUPAC Name (2S)-1-methoxypropan-2-ol
SMILES CC(COC)O

Specifications

Color Colorless
Boiling Point 120°C
Quantity 1 mL
UN Number 3092
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Methoxy-2-propanol
EINECSNumber (2)-0404
RTECSNumber UB7700000
TSCA No