missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Cyclohexylethylamine 98.0+%, TCI America™

Catalog No. C15315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15315G 5 g
C153125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15315G Supplier TCI America Supplier No. C15315G

Chemical Identifiers

CAS 17430-98-7
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00066261
InChI Key XBWOPGDJMAJJDG-UHFFFAOYNA-N
Synonym s-+-1-cyclohexylethylamine, s-1-cyclohexylethanamine, 1s-1-cyclohexylethanamine, s-1-cyclohexylethylamine, 1s-1-cyclohexylethan-1-amine, chembl19424, 1-cyclohexyl-ethylamine, 1-cyclohexylethanamine #, 1s-1-cyclohexylethylamine, s-1-cyclohexyl-ethylamine
PubChem CID 5325951
IUPAC Name 1-cyclohexylethan-1-amine
SMILES CC(N)C1CCCCC1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
UN Number 2734
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Cyclohexylethylamine