missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1-Chloro-2-methylbutane 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 25 mL |
| UN Number | 1107 |
| Formula Weight | 106.59 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-(+)-1-Chloro-2-methylbutane |
Sécurité et manipulation
| EINECSNumber | (2)-0066 |
| TSCA | No |
| Recommended Storage | Refrigerator |