missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-2-cyanopyrrolidine, 95%, Thermo Scientific™

Catalog No. AAH57150MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57150MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57150MD Supplier Thermo Scientific Chemicals Supplier No. H57150MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(-)-1-Boc-2-pyrrolidinecarbonitrile is used in synthesis of prolyl oligopeptidase inhibitors and is a reagent that is used in the design of potent and selective inhibitors of DPP-4 that exhibits an attractive pharmacokinetic profile as antidiabetics.

Solubility
Slightly soluble in water.

Notes
Moisture and light sensitive. Store away from light, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

Chemical Identifiers

CAS 228244-04-0
Molecular Formula C10H16N2O2
Molecular Weight (g/mol) 196.25
MDL Number MFCD01861224
InChI Key MDMSZBHMBCNYNO-UHFFFAOYNA-N
Synonym s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
PubChem CID 6951263
IUPAC Name tert-butyl (2S)-2-cyanopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C#N

Specifications

Melting Point 33°C to 36°C
Flash Point 110°C (230°F)
Quantity 250 mg
Sensitivity Light and moisture sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +105° (c=1 in Chloroform)
Formula Weight 196.25
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Boc-2-cyanopyrrolidine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon