missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydroisoquinolylmethan-3-ol, 98%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 334730010

Quantity 1g
Packaging Glass bottle

Catalog No. AC334730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

 

Specifications

Chemical Identifiers

18881-17-9
163.22
ZSKDXMLMMQFHGW-JTQLQIEISA-N
776757
C1C(NCC2=CC=CC=C21)CO
C10H13NO
MFCD01631316
s-1,2,3,4-tetrahydroisoquinoline-3-methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylmethanol, s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, chembl61489, s-1,2,3,4-tetrahydroisoquinolin-3-yl-methanol, s---1,2,3,4-tetrahydro-3-isoquinolinemethanol, s-3-hydroxymethyl-1,2,3,4-tetrahydroisoquinoline, 3-isoquinolinemethanol, 1,2,3,4-tetrahydro-, s, s-1,2,3,4-tetrahydroisoquinolylmethan-3-ol
[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]methanol

Specifications

(S)-1,2,3,4-Tetrahydroisoquinolylmethan-3-ol
97.5
Authentic
Glass bottle
1g
-94° to 100° (20°C, 589nm) (c=1, CH3OH)
ZSKDXMLMMQFHGW-JTQLQIEISA-N
[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]methanol
776757
98%
18881-17-9
100.0
97.5% min
C10H13NO
MFCD01631316
s-1,2,3,4-tetrahydroisoquinoline-3-methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylmethanol, s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, 3s-1,2,3,4-tetrahydroisoquinolin-3-yl methanol, chembl61489, s-1,2,3,4-tetrahydroisoquinolin-3-yl-methanol, s---1,2,3,4-tetrahydro-3-isoquinolinemethanol, s-3-hydroxymethyl-1,2,3,4-tetrahydroisoquinoline, 3-isoquinolinemethanol, 1,2,3,4-tetrahydro-, s, s-1,2,3,4-tetrahydroisoquinolylmethan-3-ol
C1C(NCC2=CC=CC=C21)CO
163.22
163.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions