missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(1-Naphthyl)ethylamine, 99%

Catalog No. AAA1717606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1717606 5 g
AAA1717603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1717606 Supplier Thermo Scientific Chemicals Supplier No. A1717606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10420-89-0 | C12H14N | 172.25 g/mol

(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

Solubility
Soluble in chloroform, ethanol.

Notes
Air Sensitive. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates.

Chemical Identifiers

CAS 10420-89-0
Molecular Formula C12H14N
Molecular Weight (g/mol) 172.25
MDL Number MFCD00064179
InChI Key RTCUCQWIICFPOD-VIFPVBQESA-O
Synonym s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
PubChem CID 66325
IUPAC Name (1S)-1-naphthalen-1-ylethanamine
SMILES C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.06
Boiling Point 152°C to 153°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.623
Quantity 5 g
UN Number UN2922
Beilstein 2208024
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,ethanol.
Formula Weight 171.24
Percent Purity 99%
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
Show More Show Less
Hazard Category H301-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only