missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RXRA Recombinant Rabbit Monoclonal Antibody (23GB2815)
GREENER_CHOICE

Numéro de catalogue. MA553566
Modifier l'affichage
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA553566 100 μL
MA553567 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553566 Fournisseur Invitrogen™ Code fournisseur MA553566
Il en reste null

Rabbit Recombinant Monoclonal Antibody

RXRA Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.

Spécifications

Antigen RXRA
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2815
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rxra
Gene Accession No. P19793, P28700, Q05343
Gene Alias 9530071D11Rik; Nr2b1; Nuclear receptor subfamily 2 group B member 1; retinoic acid receptor RXR-alpha; retinoid X nuclear receptor alpha; retinoid X receptor; retinoid X receptor alpha; retinoid X receptor, alpha; rxr alpha; RXR alpha 1; Rxra; RXRalpha1
Gene Symbols Rxra
Host Species Rabbit
Immunogen A synthesized peptide derived from human Retinoid X Receptor alpha (1-41AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20181, 25271, 6256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats