missing translation for 'onlineSavingsMsg'
Learn More

(+)-Rutin trihydrate, 95%

Catalog No. AAA1357014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357014 25 g
AAA1357022 100 g
AAA1357036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1357014 Supplier Thermo Scientific Chemicals Supplier No. A1357014

CAS: 250249-75-3 | C27H36O19 | 664.566 g/mol

Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

Solubility
Soluble in DMSO, acetone, dichloromethane or ethyl acetate. Insoluble in water.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 250249-75-3
Molecular Formula C27H36O19
Molecular Weight (g/mol) 664.566
MDL Number MFCD00149490
InChI Key NLLBWFFSGHKUSY-JPRRWYCFSA-N
Synonym rutoside trihydrate, +-rutin trihydrate, rutin trihydrate, rutin hydrate, unii-rf4n03853g, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-3-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyltetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy-4h-chromen-4-one trihydrate, rutoside trihydrate ep, rutin hydrate jan, rutin trihydrate hplc, rutin trihydrate, analytical standard
PubChem CID 16218542
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)O)C5=CC(=C(C=C5)O)O)O)O)O)O)O)O.O.O.O

Specifications

Melting Point 184°C (decomposition)
Quantity 25 g
Beilstein 75455
Sensitivity Light sensitive
Merck Index 14,3802
Solubility Information Soluble in DMSO,acetone,dichloromethane or ethyl acetate. Insoluble in water.
Optical Rotation +9° (c=0.5 in Ethanol)
Formula Weight 664.58 (610.52 Anhydrous)
Percent Purity 97%
Chemical Name or Material (+)-Rutin trihydrate
EINECSNumber 205-814-1
RTECSNumber VM2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only