missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLRU32X

 View more versions of this product

Catalog No. PLRU3-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ru1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Ruthenium Standard
7440-18-8,7647-01-0,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,10.0,89.9
MFCD00011207 MFCD03458417
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
KJTLSVCANCCWHF-UHFFFAOYSA-N
10% HCl
101.07
23950
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
0.1,10.0,89.9
Ru
AA, ICP
Certified Reference Material
[Ru]
ruthenium
500 mL
CHEBI:30682
Matrix    Concentration    Cat. No.
10% HCl    1000μg/mL    PLRU3-2Y, -2X    10,000    PLRU3-3Y, -3X

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
CHEBI:30682
[Ru]
Ru
MFCD00011207 MFCD03458417
23950
ruthenium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only