CAS RN 7440-18-8
Ruthenium standard solution, for AAS, 1mg/mL Ru in 10-20% HCl, Thermo Scientific Chemicals
CAS: 7440-18-8 | Molecular Formula: Ru | Molecular Weight: 101.07 g/mol
Ruthenium, 10% on activated carbon powder, reduced
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 0.5% on granular carbon
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, plasma standard solution, Specpure™ Ru 10,000μg/mL
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 2% on 3.18mm (0.125in) alumina pellets
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium powder, -200 mesh, Premion™, 99.99+% (metals basis excluding Ca), Ca 100ppm max
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium powder, -325 mesh, 99.9% (metals basis)
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium powder, -200 mesh, 99.95% (metals basis)
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium black, 99.9% (metals basis)
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium cubes, 6mm (0.24in) square, 99.9% (metals basis)
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 5% on activated carbon powder, reduced
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium, 5% on alumina powder, Thermo Scientific Chemicals
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
Ruthenium 5% on Carbon (wetted with ca. 50% Water), TCI America™
CAS: 7440-18-8 Molecular Formula: Ru Molecular Weight (g/mol): 101.07 MDL Number: MFCD00011207 MFCD03458417 InChI Key: KJTLSVCANCCWHF-UHFFFAOYSA-N Synonym: on carbon,rutenio,black,unii-7ui0tkc3u5,7ui0tkc3u5,ruthenium, powder,alumina,atom,rutherium black,ruthenium/carbon PubChem CID: 23950 ChEBI: CHEBI:30682 IUPAC Name: ruthenium SMILES: [Ru]
- 1
- 2