missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$685.00 - $1155.63
Chemical Identifiers
| CAS | 7440-18-8 |
|---|---|
| Molecular Formula | Ru |
| Molecular Weight (g/mol) | 101.07 |
| MDL Number | MFCD00011207 MFCD03458417 |
| InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
| PubChem CID | 23950 |
| ChEBI | CHEBI:30682 |
| IUPAC Name | ruthenium |
| SMILES | [Ru] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLRU3-2Y
|
SPEX CertiPrep Group LLC
PLRU32Y |
125 mL | Triple-leached LDPE bottle |
Each for $685.00
|
|
||||
|
PLRU3-2X
|
SPEX CertiPrep Group LLC
PLRU32X |
500 mL | Triple-leached LDPE bottle |
Each for $1,155.63
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ru | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-18-8 | |
| 101.07 | |
| KJTLSVCANCCWHF-UHFFFAOYSA-N | |
| CHEBI:30682 | |
| [Ru] |
| Ru | |
| MFCD00011207 MFCD03458417 | |
| 23950 | |
| ruthenium |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 0.1,10.0,89.9 | |
| Ru | |
| AA, ICP | |
| Certified Reference Material | |
| [Ru] | |
| ruthenium | |
| 125 mL | |
| CHEBI:30682 | |
| Matrix Concentration Cat. No. 10% HCl 1000μg/mL PLRU3-2Y, -2X 10,000 PLRU3-3Y, -3X |
| 7440-18-8,7647-01-0,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,10.0,89.9 | |
| MFCD00011207 MFCD03458417 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| KJTLSVCANCCWHF-UHFFFAOYSA-N | |
| 10% HCl | |
| 101.07 | |
| 23950 | |
| Assurance | |
| Ruthenium Standard |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only