missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) 2,4-pentanedionate, Ru 24% min

Catalog No. AA1056803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1056803 1 g
AA1056806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1056803 Supplier Thermo Scientific Chemicals Supplier No. 01056803
Only null left

For Hydrogenation | CAS: 14284-93-6 | C15H21O6Ru | 398.40 g/mol

Precursor for the preparation of other compounds of ruthenium. Ruthenium(III) 2,4-pentanedionate is employed as a recyclable catalyst for certain organic transformations like the acetylation of phenols, alcohols, and amines under neat conditions. It is also used as a homogeneous catalyst, for example, in the hydrolysis of sodium borohydride and regiospecific tritiation of aromatic carboxylic acids. It is also used as a catalyst for enantioselective hydrogenation of acids such as aryl acrylic acid and aryl propenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14284-93-6
Molecular Formula C15H21O6Ru
Molecular Weight (g/mol) 398.40
MDL Number MFCD00000030
InChI Key RTZYCRSRNSTRGC-LNTINUHCSA-K
Synonym ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+
PubChem CID 5488829
SMILES [Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C (decomposition)
Color White
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in most organic solvents such as acetone,chlorinated hydrocarbons,alcohols,cyclohexane and benzene. Insoluble in water.
IUPAC Name (Z)-4-oxopent-2-en-2-olate;ruthenium(3+)
Formula Weight 398.4
Appearance Red crystals
Chemical Name or Material Ruthenium(III) 2, 4-pentanedionate, Ru 24% min.
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-193-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only