missing translation for 'onlineSavingsMsg'
Learn More

RuCl2[(S)-xylbinap][(S,S)-dpen], TCI America™

Catalog No. p-7135717
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0134200MG 200 mg
R01341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0134200MG Supplier TCI America Supplier No. R0134200MG

Chemical Identifiers

CAS 220114-03-4
Molecular Formula C66H64Cl2N2P2Ru
Molecular Weight (g/mol) 1119.17
MDL Number MFCD09753030
InChI Key HYMSONXJNGZZBM-LISIALKWSA-L
Synonym Dichloro[(S)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 11136945
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1S,2S)-1,2-diphenylethane-1,2-diamine
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl

Specifications

Quantity 200 mg
Formula Weight 1119.17
Physical Form Crystalline Powder
Chemical Name or Material RuCl2[(S)-xylbinap][(S,S)-dpen]