missing translation for 'onlineSavingsMsg'
Learn More

RuCl[(S,S)-Tsdpen](p-cymene) 90.0+%, TCI America™

Catalog No. R0124200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0124200MG 200 mg
R01241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0124200MG Supplier TCI America Supplier No. R0124200MG
Only null left

Chemical Identifiers

CAS 192139-90-5
Molecular Formula C31H35ClN2O2RuS
MDL Number MFCD09953484
Synonym [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)

Specifications

Quantity 200 mg
Formula Weight 636.21
Percent Purity ≥90% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material RuCl[(S,S)-Tsdpen](p-cymene)