missing translation for 'onlineSavingsMsg'
Learn More

[RuCl(p-cymene)((S)-xylbinap)]Cl, TCI America™

Supplier:  TCI America R01511G

 View more versions of this product

Catalog No. R01511G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

[RuCl(p-cymene)((S)-xylbinap)]Cl
C62H62Cl2P2Ru
1 g
CBZXDZCPCDMKJU-UHFFFAOYSA-L
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
72376324
Crystalline Powder
944451-25-6
MFCD09753015
Chloro[(S)-(-)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.Cl[Ru]Cl
1041.096
1041.10

Chemical Identifiers

944451-25-6
1041.096
CBZXDZCPCDMKJU-UHFFFAOYSA-L
72376324
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.Cl[Ru]Cl
C62H62Cl2P2Ru
MFCD09753015
Chloro[(S)-(-)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions