missing translation for 'onlineSavingsMsg'
Learn More

[RuCl(p-cymene)((S)-xylbinap)]Cl, TCI America™

Catalog No. p-7135730
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0151200MG 200 mg
R01511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0151200MG Supplier TCI America Supplier No. R0151200MG
Only null left

Chemical Identifiers

CAS 944451-25-6
Molecular Formula C62H62Cl2P2Ru
Molecular Weight (g/mol) 1041.096
MDL Number MFCD09753015
InChI Key CBZXDZCPCDMKJU-UHFFFAOYSA-L
Synonym Chloro[(S)-(-)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
PubChem CID 72376324
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.Cl[Ru]Cl

Specifications

Quantity 200 mg
Formula Weight 1041.10
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((S)-xylbinap)]Cl