accessibility menu, dialog, popup

UserName

CH3CH3H3CCH3PPCH3H3CH3CCH3CH3H3CH3CClRuCl(?)

CAS RN 944451-25-6

IUPAC Name: 1-methyl-4-(propan-2-yl)benzene; dichlororuthenium; {2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane
Synonyms: cymene; dichlororuthenium; {2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 1041.1
Molecular Formula: C62H62Cl2P2Ru
InChi Key: CBZXDZCPCDMKJU-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results