missing translation for 'onlineSavingsMsg'
Learn More

Rubidium ICP Standard, 1000 ppm Rb in 3% HNO3, Ricca Chemical

Catalog No. PRB1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRB1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRB1KN100 Supplier Ricca Chemical Company Supplier No. PRB1KN100
Only null left

Chemical Identifiers

CAS 13126-12-0 , 7697-37-2 , 7732-18-5
Molecular Formula NO3Rb
Molecular Weight (g/mol) 147.47
MDL Number MFCD00011193
InChI Key RTHYXYOJKHGZJT-UHFFFAOYSA-N
PubChem CID 25731
IUPAC Name rubidium(1+) nitrate
SMILES [Rb+].[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm
CAS Min % 94.0, 4.22, 0.16
CAS Max % 97.0, 4.39, 0.17
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Solvent or Matrix 3% HNO
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Rubidium ICP Standard
Show More Show Less