missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium AA Standard, 1000 ppm Rb in 3% HNO3, Ricca Chemical
Chemical Identifiers
| CAS | 13126-12-0 , 7697-37-2 , 7732-18-5 |
|---|---|
| Molecular Formula | NO3Rb |
| Molecular Weight (g/mol) | 147.47 |
| MDL Number | MFCD00011193 |
| InChI Key | RTHYXYOJKHGZJT-UHFFFAOYSA-N |
| PubChem CID | 25731 |
| IUPAC Name | rubidium(1+) nitrate |
| SMILES | [Rb+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|
ARB1KN100
|
N/A | Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
ARB1KN500
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 13126-12-0 , 7697-37-2 , 7732-18-5 | |
| 147.47 | |
| RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
| rubidium(1+) nitrate |
| NO3Rb | |
| MFCD00011193 | |
| 25731 | |
| [Rb+].[O-][N+]([O-])=O |
Specifications
| 13126-12-0 , 7697-37-2 , 7732-18-5 | |
| 97.0, 4.39, 0.17 | |
| MFCD00011193 | |
| [Rb+].[O-][N+]([O-])=O | |
| 147.47 | |
| 1mL = 1mg Rb (1, 000ppm Rb)RbNO3 in 3% HNO3 |
| 94.0, 4.22, 0.16 | |
| NO3Rb | |
| RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
| rubidium(1+) nitrate | |
| 25731 | |
| Rubidium AA Standard |
Safety and Handling
ShelfLife : 24 months