missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium AA Standard, 1000 ppm Rb in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company ARB1KN500
| Packaging | Natural Poly Bottle |
|---|
Chemical Identifiers
| 13126-12-0 , 7697-37-2 , 7732-18-5 | |
| 147.47 | |
| RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
| rubidium(1+) nitrate |
| NO3Rb | |
| MFCD00011193 | |
| 25731 | |
| [Rb+].[O-][N+]([O-])=O |
Specifications
| Rubidium AA Standard | |
| 13126-12-0 , 7697-37-2 , 7732-18-5 | |
| 97.0, 4.39, 0.17 | |
| MFCD00011193 | |
| [Rb+].[O-][N+]([O-])=O | |
| 147.47 | |
| 1mL = 1mg Rb (1, 000ppm Rb)RbNO3 in 3% HNO3 |
| Natural Poly Bottle | |
| 94.0, 4.22, 0.16 | |
| NO3Rb | |
| RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
| rubidium(1+) nitrate | |
| 25731 |
Safety and Handling
ShelfLife : 24 months