missing translation for 'onlineSavingsMsg'
Learn More

Rubidium AA Standard, 1000 ppm Rb in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company ARB1KN100

Packaging Natural LDPE Bottle

 View more versions of this product

Catalog No. ARB1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

13126-12-0 , 7697-37-2 , 7732-18-5
147.47
RTHYXYOJKHGZJT-UHFFFAOYSA-N
rubidium(1+) nitrate
NO3Rb
MFCD00011193
25731
[Rb+].[O-][N+]([O-])=O

Specifications

Rubidium AA Standard
13126-12-0 , 7697-37-2 , 7732-18-5
97.0, 4.39, 0.17
MFCD00011193
[Rb+].[O-][N+]([O-])=O
147.47
1mL = 1mg Rb (1, 000ppm Rb)RbNO3 in 3% HNO3
Natural LDPE Bottle
94.0, 4.22, 0.16
NO3Rb
RTHYXYOJKHGZJT-UHFFFAOYSA-N
rubidium(1+) nitrate
25731
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents

Promotions

Promotions