accessibility menu, dialog, popup

UserName

CAS RN 13126-12-0

RbONOO

CAS RN 13126-12-0

IUPAC Name: rubidium(1+) nitrate
Synonyms: rubidium nitrate
Molecular Weight (g/mol): 147.47
Molecular Formula: NO3Rb
InChi Key: RTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES: [Rb+].[O-][N+]([O-])=O
Grade 
  • (8)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (8)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)