missing translation for 'onlineSavingsMsg'
Learn More

Rubeanic Acid 98.0+%, TCI America™

Catalog No. D09575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09575G Supplier TCI America Supplier No. D09575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-40-3
Molecular Formula C2H4N2S2
Molecular Weight (g/mol) 120.188
MDL Number MFCD00004941
InChI Key OAEGRYMCJYIXQT-UHFFFAOYSA-N
Synonym dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6
PubChem CID 2777982
IUPAC Name ethanedithioamide
SMILES C(=S)(C(=S)N)N

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 120.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Rubeanic Acid
RTECSNumber RP1575000
TSCA Yes