missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ru-MACHO(regR)-BH, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 586.42 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ru-MACHO(regR)-BH |
Safety and Handling
| TSCA | No |