missing translation for 'onlineSavingsMsg'
Learn More

(RS)-(Methylenecyclopropyl)acetic acid, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012410
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012410 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012410 Supplier Merck Emd Millipore Supplier No. 63978100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages

Specifications

CAS 1073-00-3
Physical Form Neat
Linear Formula C6H8O2
MDL Number MFCD11558999
Type Analytical Standard
Quantity 100 mg
Formula Weight 112.13
Percent Purity ≥95% (HPLC)
Grade Analytical Standard