missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPI 1 95.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 50 mg |
| Formula Weight | 297.31 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | RPI 1 |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |