missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPH3A Polyclonal Antibody

Description
PA1-774 detects rabphilin 3A from rat tissues. PA1-774 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~75 kDa protein representing rabphilin 3A from rat brain whole protein extract. PA1-774 immunogen is a synthetic peptide corresponding to residues M(1) T D T V V N R W M Y P G D G P L Q(18) C of rat Rabphilin 3A. This sequence is completely conserved between rat and mouse rabphilin 3A. PA1-774 immunizing peptide (Cat. # PEP-102) is available for use in neutralization and control experiments.
The carboxy-terminal of rabphilin-3A consists of two C2 domains, A and B, and the amino-terminal (residues 45-170) contains a cysteine-rich rich region with two zinc finger motifs. Rabphilin-3A belongs to a family of other carboxyterminal type (C-type) tandem C2 proteins, which includes synaptotagmins and Doc2. Rabphilin is expressed in neuroendocrine cells and co-localizes with Rab3A on synaptic vesicles and chromaffin granules. Rabphilin-3A binds Rab3a/GTP/Mg+ within amino-terminal residues 45 and 170. Rabphilin-3A binds calcium ions and phosphatidylinositol 4,5-bisphophate containing lipid vesicles within its C2 domains. Rabphilin-3A is a positive regulator of calcium dependent exocytosis, while Rab3a is a negative regulator of exocytosis. Although rabphilin-3A associates with Rab3a, they seem to influence exocytosis independently of each other. Rabphilin-3A effects are likely mediated through interactions with an unknown factor that recognizes the Rab3 binding domain.
Spécifications
Spécifications
| Antigen | RPH3A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | RPH3A |
| Gene Accession No. | P47708, P47709 |
| Gene Alias | 2900002P20Rik; AU022689; AW108370; Exophilin 1; Exophilin1; Exophilin-1; KIAA0985; rabphilin 3A; rabphilin 3A homolog; rabphilin-3A; RPH3A |
| Gene Symbols | RPH3A |
| Afficher plus de résultats |