missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPGRIP1 Monoclonal Antibody (5H2)
GREENER_CHOICE

Numéro de catalogue. PIMA524480
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIMA524480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIMA524480 Fournisseur Invitrogen™ Code fournisseur MA524480
Il en reste null

Mouse Monoclonal Antibody

Peptide Sequence: QGRRRFLFDM LNGQDPDQGH LKFTVVSDPL DEEKKECEEV GYAYLQLWQI LESGRDILEQ ELDIVSPEDL ATPIGRLKVS LQAAAVLHAI YKEMTEDLFS.

May function as scaffolding protein. Required for normal location of RPGR at the connecting cilium of photoreceptor cells. Required for normal disk morphogenesis and disk organization in the outer segment of photoreceptor cells and for survival of photoreceptor cells.

Spécifications

Antigen RPGRIP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RPGRIP1
Gene Accession No. Q96KN7
Gene Alias 0610005A07Rik; 4930401L23Rik; 4930505G06Rik; A930002K18Rik; AA415034; CORD13; LCA6; nmf247; retinitis pigmentosa GTPase regulator interacting protein 1; RGI1; RGRIP; RPGR-interacting protein 1; RPGRIP; Rpgrip1; RPGRIP1d; X-linked retinitis pigmentosa GTPase regulator-interacting protein 1
Gene Symbols RPGRIP1
Host Species Mouse
Immunogen RPGRIP1 (NP_065099.2, 1187 a.a. ~ 1286 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57096
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats