missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA43/TWISTNB Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
Spécifications
Spécifications
| Antigène | RPA43/TWISTNB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | TWISTNB |
| Numéro d’acquisition du gène | Q3B726 |
| Alias du gène | DNA-directed RNA polymerase I subunit RPA43, twist neighbor protein |
| Symboles de gène | TWISTNB |
| Afficher plus de résultats |