missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA32 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
Spécifications
Spécifications
| Antigène | RPA32 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | RPA2 |
| Numéro d’acquisition du gène | P15927, Q62193 |
| Alias du gène | REPA2, RPA32 |
| Symboles de gène | RPA2 |
| Afficher plus de résultats |