Learn More
Thermo Scientific™ RP6 Windshielded Swinging Bucket Rotor
Catalog No. 11178119
Change view
Click to view available options
Capacity:
Not Applicable
Description:
RP6 Windshield Rotor Lid (required)
Catalog No. Capacity Description
11178119 Not Applicable RP6 Windshield Rotor Lid (required)
1 options
Catalog No. 11178119 Supplier Thermo Scientific™ Supplier No. 11178119
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
  • Capacity: 6 × 1000mL (6L) with Round bucket, 12 × 250mL (3L) with Rectangular bucket
  • Max. speed/RCF: 4250rpm/5,735 x g with Round bucket, 5,392 x g with Rectangular rotor
Ordering Information:
Order rotor body (11178252) and buckets and inserts to complete assembly. Order 11178216 for rectangular bucket sealing cap and 25144058 for round bucket sealing cap. Contact your Sales Representative for information.
Includes:
Rotor body only
Compatible with:
Thermo Scientific KR4i and Sorvall HT6 Centrifuges
Specifications
Description RP6 Windshield Rotor Lid (required)
Type Bucket Lid
Capacity Not Applicable
For Use With (Equipment) Thermo Scientific Sorvall HT6 and KR4i Large Capacity Centrifuges
Biocontainment Yes
Quantity 1