missing translation for 'onlineSavingsMsg'
Learn More

Rotenone, ≥97%, MP Biomedicals™

Catalog No. ICN15015405
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15015405 5 g
ICN15015410 10 g
ICN15015425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15015405 Supplier MP Biomedicals Inc Supplier No. 0215015405

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83-79-4
Molecular Formula C23H22O6
Molecular Weight (g/mol) 394.423
InChI Key JUVIOZPCNVVQFO-HBGVWJBISA-N
Synonym rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
PubChem CID 6758
ChEBI CHEBI:28201
SMILES CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC

Specifications

Color Beige to White
Assay Percent Range ≥97%
Quantity 5 g
Formula Weight 394.4
Physical Form Powder
Chemical Name or Material Rotenone