missing translation for 'onlineSavingsMsg'
Learn More

Ropinirole Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018605
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018605 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018605 Supplier Merck Emd Millipore Supplier No. PHR1633200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 91374-20-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H24N2 O · HCl
MDL Number MFCD01754173
Synonym Ropinirole hydrochloride; 4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2 H-indol-2-one hydrochloride; SKF 101468 hydrochloride
Type Certified Reference Material
Quantity 200 mg
Formula Weight 296.84
Grade Certified Reference Material