missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Romifidine 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White-Yellow |
| Quantity | 50 mg |
| Formula Weight | 258.09 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Romifidine |
Safety and Handling
| TSCA | No |