missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rochelle Salt Stabilizer, for Ammonia Determination, Ricca Chemical
$61.75 - $349.47
Specifications
| CAS | 6381-59-5,7732-18-5 |
|---|---|
| CAS Min % | 65.0,32.68 |
| CAS Max % | 68.0,34.01 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |