missing translation for 'onlineSavingsMsg'
Learn More

Rochelle Salt Stabilizer, for Ammonia Determination, Ricca Chemical

Catalog No. 66404
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66404 120 mL
664016 500 mL
664032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 66404 Supplier Ricca Chemical Company Supplier No. 66404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Rochelle Salt Stabilizer
Color Colorless
Name Note For Ammonia Determination
CAS 6381-59-5, 7732-18-5
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Molecular Formula C4H12KNaO10
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Molecular Weight (g/mol) 282.218
PubChem CID 2724148
Grade Laboratory
Packaging Natural Poly Bottle
pH 7 to 8
Quantity 120 mL
Physical Form Liquid
Show More Show Less