missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rochelle Salt Stabilizer, for Ammonia Determination, Ricca Chemical
Supplier: Ricca Chemical Company 664032
Specifications
| Rochelle Salt Stabilizer | |
| For Ammonia Determination | |
| 65.0,32.68 | |
| 282.22 g/mol | |
| Natural Poly Bottle | |
| 1 L |
| Colorless | |
| 6381-59-5,7732-18-5 | |
| 68.0,34.01 | |
| Laboratory | |
| 7 to 8 | |
| Liquid |