Learn More
Roche Diagnostics Cholesterol Test Strips for Accutrend Plus Meters
Catalog No. 22045730
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
22045730 Each
1 options
Catalog No. 22045730 Supplier Roche Diagnostics Supplier No. BMC11418262171
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cholesterol results in three minutes
  • Can be dosed outside of the meter to prevent cross-contamination for community health screenings
Specifications
Content And Storage Room Temperature
Precision LO= 2.9%, x= 203, range 173 to 224mg/dL
Quantity Each
Range 150 to 300mg/dL
Sample Size 15 to 40 μL
Test Time 180 sec.