missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Robotic aTIP 30μL Filter Pipette Tips
GREENER_CHOICE

Numéro de catalogue. 12111063
Change view
Click to view available options
Volume (Metric):
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric)
12111063 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 12111063 Fournisseur BIOTIX™ Code fournisseur 63300183
Il en reste null

For use with Agilent™ Velocity 11™ VPrep and Bravo, 384-Head platforms.

  • Made from clear polypropylene
  • Compatible with 96-and 384-well plates

Spécifications

Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Format Rack
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Agilent/Velocity 11 VPrep and Bravo, 384 Tip Format
Graduated No
Material Polypropylene
Shape Conical
Afficher plus de résultats Afficher moins de résultats