missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robert's Reagent, for the Qualitative Detection of Albumin in Urine, Ricca Chemical
Qualitative Detection of Albumin in Urine
$298.40 - $298.40
Specifications
| CAS | 7732-18-5 , 10034-99-8 , 7697-37-2 |
|---|---|
| CAS Min % | 57.0,27.19,14.21 |
| CAS Max % | 59.0,28.3,14.79 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
Specifications
| 7732-18-5 , 10034-99-8 , 7697-37-2 | |
| 59.0,28.3,14.79 | |
| Amber Glass Bottle |
| 57.0,27.19,14.21 | |
| Laboratory |