Research Products Corporation RNase Away, 475ml Spray Bottle, Effective for Use with Electrophoresis Equipment, Glass Plates, Other Laboratory Equipment
Catalog No. 501978158
RNase Away removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment.
Catalog No.
501978158
Supplier
Research Products CorporationSupplier No.
RPI147003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RNase Away removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment.