Learn More
Research Products Corporation RNase Away, 4 Liter Bottle
Catalog No. 50212367
Encompass
RNase Away removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment.
Catalog No. 50212367 Supplier Research Products Corporation Supplier No. RPI 147002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RNase Away removes RNase and DNA contamination from glassware and plastic quickly and economically. Simply apply RNase away to surface or soak as needed and rinse with water. Non-abrasive formulation does not contain strong acids. Effective for use with electrophoresis equipment, glass plates, and other laboratory equipment.