missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega RNase A Solution
RNase A is an endoribonuclease that specifically hydrolyzes RNA 3' of pyrimidine residues and cleaves the phosphodiester linkage to the adjacent nucleotide. RNase A is used to remove RNA during procedures for the isolation of plasmid and genomic DNA.
$209.35 - $913.09
Specifications
| Concentration | 4mg/mL |
|---|---|
| Content And Storage | 15°C to 30°C |
| Product Type | RNase A Solution, 4mg/mL |
Description
- Remove RNA During Plasmid and Genomic DNA Isolation Procedures
- Supplied at a concentration of 4mg/mL
Specifications
| 4mg/mL | |
| 15°C to 30°C | |
| RNase A Solution, 4mg/mL |