missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega RNase A Solution
Description
Includes
Binding Buffer, ReliaPrep Resin, Wash Buffer, 10mM EDTA (pH 8.0), 25mM Tris-HCl (pH 8.0), Proteinase K (PK) Solution, RNase A Solution, Cell Lysis Buffer (CLD)
- Remove RNA During Plasmid and Genomic DNA Isolation Procedures
- Supplied at a concentration of 4mg/mL
Specifications
Specifications
| Concentration | 4mg/mL |
| Content And Storage | 15°C to 30°C |
| Format | Multiwell |
| For Use With (Equipment) | ReliaPrep™ 96 genomic DNA Miniprep HT system |
| For Use With (Application) | Rapid isolation of genomic DNA |
| Includes | Binding Buffer, ReliaPrep™ Resin, Wash Buffer, 10mM EDTA (pH 8), 25mM Tris HCl (pH 8), Proteinase K solution, RNase A solution, Cell Lysis Buffer (CLD) |
| Product Type | RNase A Solution, 4mg/mL |
| Quantity | 5 mL |