missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Control 250 with Manual

Catalog No. AM7155
Encompass
Change view
Click to view available options
Quantity:
2 x 20 μL, with Manual
2 x 20μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7155 2 x 20μL
AM7155M 2 x 20 μL, with Manual
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM7155 Supplier Invitrogen™ Supplier No. AM7155

Includes

RNA Control and THE RNA Storage Solution

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers.

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each. RNA Control 250 consists of a single, ultra-pure, 2000nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Invitrogen THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).

• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage RNA Control and THE RNA Storage Solution should be stored at –20°C
Format Tube
Label or Dye Unconjugated
Product Line Ambion
Quantity 2 x 20μL
Shipping Condition Dry Ice
Volume 2 x 20 μL
What is the sequence of RNA Control 250?

The sequence of RNA Control 250 is proprietary.

For Research Use Only. Not for use in diagnostic procedures.