missing translation for 'onlineSavingsMsg'
Learn More

Ritonavir, 98%, Thermo Scientific Chemicals

Catalog No. AC461222500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461222500 250 mg
AC461220010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461222500 Supplier Thermo Scientific Chemicals Supplier No. 461222500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155213-67-5
Molecular Formula C37H48N6O5S2
Molecular Weight (g/mol) 720.94
InChI Key NCDNCNXCDXHOMX-XGKFQTDJSA-N
Synonym ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn
PubChem CID 392622
ChEBI CHEBI:45409
IUPAC Name 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate
SMILES CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O

Specifications

Melting Point 120.0°C to 132.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: <1 mg/mL. Other solubilities: 20 mg/mL dmso, 1 mg/mL ethanol, slightly soluble in methanol, soluble in isopropanol
Formula Weight 720.94
Percent Purity 98%
Physical Form Gel
Chemical Name or Material Ritonavir
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed, in contact with skin or if inhaled

RUO – Research Use Only