missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ritodrine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 195°C |
| Color | White-Yellow |
| Quantity | 250 mg |
| Formula Weight | 323.82 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ritodrine Hydrochloride |
Safety and Handling
| RTECSNumber | DO6524000 |
| TSCA | No |