missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ritalinic acid

Catalog No. p-7214433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463000010 1 g
AC463000050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463000010 Supplier Thermo Scientific™ Supplier No. 463000010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 19395-41-6
Molecular Formula C13H17NO2
Molecular Weight (g/mol) 219.28
InChI Key INGSNVSERUZOAK-UHFFFAOYNA-N
IUPAC Name 2-phenyl-2-(piperidin-2-yl)acetic acid
SMILES OC(=O)C(C1CCCCN1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 219.28
Chemical Name or Material Ritalinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

EINECSNumber 243-020-7

RUO – Research Use Only