missing translation for 'onlineSavingsMsg'
Learn More

Risedronate Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018089
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018089 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018089 Supplier Merck Emd Millipore Supplier No. PHR207950MG
Only null left

Specifications

CAS 6419-36-9
Linear Formula C7H7 NO2 · HCl
MDL Number MFCD00012819
Melting Point 161°C to 163°C
Synonym 3-Pyridylacetic acid hydrochloride; 3-Pyridineacetic acid hydrochloride
Type Certified Reference Material
Quantity 50 mg
Formula Weight 173.60
Grade Certified Reference Material